Principaux documents
Z255912
Nalgene® 180 vacuum tubing
I.D. × O.D. 3/16 in. × 9/16 in., wall size 3/16 in.
Synonyme(s) :
plastic vacuum tubing
About This Item
Code UNSPSC :
41105108
Conditionnement
pkg of 10 feet
Fabricant/nom de marque
Nalgene 8000-0055
Paramètres
35 psi max. pressure (52 °C)
70 psi max. pressure (25 °C)
D.I. × D.E.
3/16 in. × 9/16 in.
Taille de la paroi
3/16 in.
Description générale
Transparent, nontoxic, autoclavable plastic tubing has thick walls for high vacuum.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries