Principaux documents
Z247235
Coors porcelain crucible cover
cap size D, I.D. 54 mm
1 PKG
202,00 €
202,00 €
Check Cart for Availability
Changer de vue
1 PKG
202,00 €
About This Item
Code UNSPSC :
41122200
Nomenclature NACRES :
NB.43
202,00 €
Check Cart for Availability
Taille du bouchon
D
Conditionnement
pkg of 12 ea
Fabricant/nom de marque
Coorstek 66125
D.I.
54 mm
Description générale
Coors porcelain crucible cover is glazed all over except for the rim.
Caractéristiques et avantages
  • Ensures great resistance to thermal-shock
  • Durable
  • Rigid
  • Sustained refractory qualities
Autres remarques
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
Informations légales
Coors is a trademark of Coorstek, Inc.
Produit(s) apparenté(s)
Réf. du produit
Description
Tarif
Questions
  1. Kindly describe how many pieces of lid are there in this pack.
    1 réponse
    1. This product is sold as a package of 12 each. There are 12 covers included.
      Utile ?