Principaux documents
Z215481
Rigid PTFE sleeve
joint: ST/NS 19/26
Synonyme(s) :
ptfe sleeves
1 EA
9,73 €
9,73 €
Tarif10,50 €Économiser 7 %
Check Cart for Availability
Changer de vue
1 EA
9,73 €
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
9,73 €
Tarif10,50 €Économiser 7 %
Check Cart for Availability
Joint
joint: ST/NS19/26
Fabricant/nom de marque
Cowie
Description générale
Robust, PTFE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free, high-vacuum performance at temperatures up to 280°C.