Principaux documents
Z147338
Sigma-Aldrich
Lecture-bottle adapter, NPT to NPT
stainless steel
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Matériaux
stainless steel
Entrée et sortie NPTM × NPTM
1/4 in. × 1/4 in.
Description générale
For connection of valves, regulators, and filters to cylinder valves.