Principaux documents
Z117021
Stainless steel 304 syringe needle
gauge 14, L 6 in.
1 EA
51,30 €
51,30 €
Check Cart for Availability
Changer de vue
1 EA
51,30 €
About This Item
Code UNSPSC :
41122002
Nomenclature NACRES :
NB.03
51,30 €
Check Cart for Availability
Description
Standard Hub with 304 Stainless Steel Tubing
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Cadence Science
Longueur
6 in.
Calibre
14
Catégories apparentées
Description générale
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing