Principaux documents
Z101931
Sigma-Aldrich
Single-bulb ball-tube
capacity 1,000 mL, joint: ST/NS 24/40, plain
1 EA
193,00 €
193,00 €
Check Cart for Availability
Changer de vue
1 EA
193,00 €
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
193,00 €
Check Cart for Availability
Joint
joint: ST/NS24/40
Capacité
1,000 mL
Catégories apparentées
Description générale
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.