Principaux documents
WHA10461400
Whatman® FP 050/0 filter holder
polysulfone, for In-line filtration 25 mm
Synonyme(s) :
Z751464
About This Item
Code UNSPSC :
41104922
Nomenclature NACRES :
NB.24
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Conditionnement
pk of 1
Fabricant/nom de marque
Whatman 10461400
Whatman Article No. 28415727 (US reference)
Description générale
The female Luer lock inlet is compatible with typical syringes. Polysulfone housing can be autoclaved with membrane in place for applications requiring a sterile filter assembly.
Caractéristiques et avantages
  • Durable polysulfone plastic construction provides moderate chemical compatibility with lighter weight than stainless steel filter holders.
  • Polysulfone filter holders are often used for clarification, venting applications, particulate removal or collection from aqueous, air or gas streams.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva