Principaux documents
WHA10442213
Whatman® glass vacuum filtration device
GV050 series, GV050/2/13 Erlenmeyer flask, NS45, 1000 mL
Synonyme(s) :
Z750883
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
borosilicate glass funnel(s) (funnel)
borosilicate glass housing
stainless steel clamp(s) (clamp)
Conditionnement
pk of 1
Fabricant/nom de marque
Whatman 10442213
Whatman Article No. 28415567 (US reference)
Contenance du flacon
1000 mL
Capacité du filtre
1000 mL
Adéquation
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
Description générale
Whatman® Vacuum Type Glass Holders
Produced from borosilicate glass and available with a choice of support screen. Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
The sintered glass support is recommended for filtration and biological analysis. The 304 stainless steel support screen is suitable for use with proteinaceous solutions.
Caractéristiques et avantages
  • Accessory to GV050 Glass Filter Funnel
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva