Principaux documents
WHA10312753
Whatman® prepleated qualitative filter paper, Grade 604 1/2
circles, diam. 320 mm, pack of 100
Synonyme(s) :
Z749281
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
cellulose filters
circles
Conditionnement
pack of 100
Fabricant/nom de marque
Whatman 10312753
Whatman Article No. 28414304 (US reference)
Paramètres
50 sec/100 mL speed (Herzberg)
Diamètre
320 mm
Dimension de pores
25 μm
Poids de base
80 g/m2
Catégories apparentées
Description générale
Grade 604½ Qualitative Filter Papers for coarse precipitates.
Cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters give improved flow rate and increased loading capacity compared to equivalent quadrant folded filters.
Caractéristiques et avantages
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.
  • Increased total loading capacity as more filter area is available.
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.
  • The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva