Principaux documents
TEM-CF300-NI-TH
Sigma-Aldrich
Carbon Film Supported Nickel Square Mesh
Thick, size 300 mesh, box of 50
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50
Taille
300 mesh
Description générale
A 10 nm thick film of pure carbon supported standard squre mesh grids. Nickel grid, 300 mesh.