Documents
TEM-76043-23
Sigma-Aldrich
Cryo-SiN TEM Windows
10 nm Thick, window size 9
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Taille de la fenêtre
9
Description générale
Silicon Nitride 10 nm thick TEM Windows (9 Windows). Ideal substrate for Cryo-Microscopy. Avoid substrate degradation. Plasma cleanable to reduce contaminants.