Principaux documents
SYNM202403
Synthware vacuum manifold with high vacuum valves
number of ports, 3, Hose Connection: right
Synonyme(s) :
Synthware M202403, vacuum manifold, vacuum unit
1 EA
486,00 €
486,00 €
Check Cart for Availability
Changer de vue
1 EA
486,00 €
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
486,00 €
Check Cart for Availability
Caractéristiques
number of ports 3
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware M202403
Description générale
With a serrated hose connection(10 mm O.D.) at right hand side. Main tube is 22 mm O.D. and the distance between valves is 100 mm center to center.
Informations légales
Synthware is a trademark of Kemtech America Inc.