Documents
SYNA232424
Synthware anti-splash adapter with fritted disc
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonyme(s) :
Synthware A232424, rotary evaporator bump trap
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS24/40
joint: ST/NS top24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A232424
Capacité
100 mL
Catégories apparentées
Description générale
With extra coarse fritted disc sealed above lower inner joint. Frit reduces foam from being carryover.
Informations légales
Synthware is a trademark of Kemtech America Inc.