Principaux documents
PL863-CGSHFBG
Plas-Labs clear perforated shelf
for PL863-CG, with bow guard, W × D × H 24 in. × 18 in. × 1/4 in.
1 EA
184,00 €
184,00 €
Check Cart for Availability
Changer de vue
1 EA
184,00 €
About This Item
Code UNSPSC :
41104510
Nomenclature NACRES :
NB.43
184,00 €
Check Cart for Availability
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Plas-Labs 863-CG/SHF/BG
Larg. × P × H
24 in. × 18 in. × 1/4 in.