Principaux documents
PL800GA-NEO-9
Plas-Labs replacement neoprene gloves
for 2 piece glove set, size 9 (for 8 in. ports)
1 PAIR
145,00 €
145,00 €
Check Cart for Availability
Changer de vue
1 PAIR
145,00 €
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
NB.35
145,00 €
Check Cart for Availability
Matériaux
neoprene
Taille
9 (for 8 in. ports)
Caractéristiques
reusable
for use with glove box sleeves
Conditionnement
pack of 1 pair
Fabricant/nom de marque
Plas-Labs 800GA-NEO-9
Description générale
Plas-Labs glovebox gloves are available as either one-piece or two-piece assemblies with interchangeable sleeves and gloves. This enables personnel to use the same glove box by changing the size or type of glove being used. Choose from four rubber types. Gloves are 15 mil thickness unless noted otherwise.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
  • White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
  • Black neoprene rubber for chemical resistance
  • Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
  • Green nitrile rubber for resistance to oil, fuel, and acids
  • Latex rubber for flexibility
Informations légales
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company