Principaux documents
MT21901297
Mettler Toledo barcode reader handheld scanner (instrument only)
1 EA
755,00 €
755,00 €
Check Cart for Availability
Changer de vue
1 EA
755,00 €
About This Item
Code UNSPSC :
41115603
Nomenclature NACRES :
NB.79
755,00 €
Check Cart for Availability
Fabricant/nom de marque
Mettler-Toledo 21901297
Description générale
General purpose corded handheld linear imager barcode reader
The following accessories are required but not suplied:
  • Cable RS232 (MT21901305)
  • Null modem adapter (MT21900924)
  • Plus one of the following:
  • Power supply unit, 5V, EU (MT21901370)
  • Power supply unit, 5V, UK (MT21901371)
Nécessaire, mais non fourni