CLS893024
Pyrex® connecting distilling adapter
joint: ST/NS 24/40
Synonyme(s) :
Corning® 8930
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.43
Produits recommandés
Joint
joint: ST/NS24/40
Fabricant/nom de marque
Corning 8930-24
D.E.
22 mm
Catégories apparentées
Description générale
The connecting arm is approximately at a 75° angle from vertical and is 200mm long.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.