Principaux documents
CAD4854
Stainless steel 304 syringe needle
gauge 18, L 10 in.
12 EA
458,00 €
458,00 €
Date d'expédition estimée le24 avril 2025
Changer de vue
12 EA
458,00 €
About This Item
Code UNSPSC :
42142600
Nomenclature NACRES :
NB.03
458,00 €
Date d'expédition estimée le24 avril 2025
Description
Standard Hub with 304 Stainless Steel Tubing
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Cadence Science 4854
Longueur
10 in.
Calibre
18
Description générale
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing