Principaux documents
BR4640040
BRAND® Dispensette® S Trace Analysis Analog-adjustable bottle-top dispenser
spring platinum-iridium, volume range 1-10 mL, without recirculation valve
Synonyme(s) :
Adjustable bottle-top dispenser, BRAND Dispensette S
1 EA
814,00 €
814,00 €
Check Cart for Availability
Changer de vue
1 EA
814,00 €
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
814,00 €
Check Cart for Availability
Caractéristiques
incremental 0.2 mL
without recirculation valve
Fabricant/nom de marque
BRAND 4640040
Gamme de volumes
1-10 mL
Raccords
(GL 28/S 28)
45 mm
GL 32
S 40
Adéquation
suitable for (suitable for HF)
Catégories apparentées
Description générale
Dispensette S Trace Analysis is specialized for dispensing high-purity acids, bases and H2O2.
Also suitable for dispensing of hydrofluoric acid when using platinum-iridium valve spring.
Caractéristiques et avantages
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Autres remarques
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Protocoles
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Contenu apparenté
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.