Principaux documents
BAF424002041
Scienceware® Lab Companion round vacuum desiccator
volume 6 L, amber polycarbonate (UV), pack of 1 ea
Synonyme(s) :
Desiccator, Desiccator cabinet, Vacuum Bell Jar, Vacuum Desiccator
1 EA
496,00 €
496,00 €
Check Cart for Availability
Changer de vue
1 EA
496,00 €
About This Item
Code UNSPSC :
41104510
Nomenclature NACRES :
NB.43
496,00 €
Check Cart for Availability
Matériaux
amber polycarbonate (UV)
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F42400-2041
Volume
6 L
6000 mL
Description générale
These high-quality vacuum desiccators provide vacuum integrity lasting three times longer than the average vacuum desiccator resulting in lower maintenance and greater peace of mind. Able to maintain 29 in.-Hg vacuum for over 72 hours at room temperature, they stay gastight allowing reliable storage or experiments over extended periods. A Viton® 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange.
Transparent polycarbonate construction is durable, easy-to-clean, chemical-resistant, and allows quick visualization of the desiccator contents. Amber tinted UV blocking models block 100% of UVA, B, and C light to minimize damages or discoloration of light-sensitive samples.
Available in both cabinet and round styles, all models include a desiccant tray (desiccants sold separately).
  • Vacuum level checking is simplified with a built-in analog vacuum gauge at the top of the unit
  • High-quality silicone O-ring and specially designed locking ring allow outstanding vacuum capability with virtually no leakage
  • Locking ring secures top and bottom halves when vacuum not in use; provides additional convenience and safety when moving the unit
  • Six models to choose from; 6, 10, and 20 L internal volume in clear or UV blocking amber
Informations légales
Lab Companion is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company