- Vanillin and 4-hydroxybenzyl alcohol promotes cell proliferation and neuroblast differentiation in the dentate gyrus of mice via the increase of brain-derived neurotrophic factor and tropomyosin-related kinase B.
Vanillin and 4-hydroxybenzyl alcohol promotes cell proliferation and neuroblast differentiation in the dentate gyrus of mice via the increase of brain-derived neurotrophic factor and tropomyosin-related kinase B.
4-Hydroxy‑3-methoxybenzaldehyde (vanillin) and 4-hydroxybenzyl alcohol (4-HBA) are well‑known phenolic compounds, which possess various therapeutic properties and are widely found in a variety of plants. In the present study, the effects of vanillin and 4‑HBA were first investigated on cell proliferation, as well as neuronal differentiation and integration of granule cells in the dentate gyrus (DG) of adolescent mice using Ki‑67, doublecortin (DCX) immunohistochemistry and 5‑bromo‑2'‑deoxyuridine (BrdU)/feminizing Locus on X 3 (NeuN) double immunofluorescence. In both the vanillin and 4‑HBA groups, the number of Ki‑67+ cells, DCX+ neuroblasts and BrdU+/NeuN+ neurons were significantly increased in the subgranular zone of the DG, as compared with the vehicle group. In addition, the levels of brain‑derived neurotrophic factor (BDNF) and tropomyosin‑related kinase B (TrkB), a BDNF receptor, were significantly increased in the DG in the vanillin and 4‑HBA groups compared with the vehicle group. These results indicated that vanillin and 4‑HBA enhanced cell proliferation, neuroblast differentiation and integration of granule cells in the DG of adolescent mice . These neurogenic effects of vanillin and 4‑HBA may be closely associated with increases in BDNF and TrkB.