Merck
  • Absorption Mechanism of a Physical Complex of Monomeric Insulin and Deoxycholyl-l-lysyl-methylester in the Small Intestine.
Absorption Mechanism of a Physical Complex of Monomeric Insulin and Deoxycholyl-l-lysyl-methylester in the Small Intestine.
Molecular pharmaceutics (2015-04-22)
Foyez Mahmud, Ok-Cheol Jeon, Taslim A Al-Hilal, Seho Kweon, Victor C Yang, Dong Soo Lee, Youngro Byun
ABSTRACT
Currently, oral administration of insulin still remains the best option to avoid the burden of repeated subcutaneous injections and to improve its pharmacokinetics. The objective of the present investigation was to demonstrate the absorption mechanism of insulin in the physical complexation of deoxycholyl-l-lysyl-methylester (DCK) for oral delivery. The oral insulin/DCK complex was prepared by making a physical complex of insulin aspart with DCK through ion-pair interaction in water. For the cellular uptake study, fluorescein-labeled insulin or DCK were prepared according to a standard protocol and applied to Caco-2 or MDCK cell lines. For the PK/PD studies, we performed intrajejunal administration of different formulation of insulin/DCK complex to diabetic rats. The resulting insulin and DCK complex demonstrated greatly enhanced lipophilicity as well as increased permeation across Caco-2 monolayers. The immunofluorescence study revealed the distribution of the complex in the cytoplasm of Caco-2 cells. Moreover, in the apical sodium bile acid transporter (ASBT) transfected MDCK, the insulin/DCK complex showed interaction with ASBT, and also demonstrated absorption through passive diffusion. We could not find that any evidence of endocytosis in relation to the uptake of insulin complex in vitro. In the rat intestine model, the highest absorption of insulin complex was observed in the jejunum at 1 h and then in the ileum at 2-4 h. In PK/PD study, the complex showed a similar PK profile to that of SC insulin. Overall, the study showed that the effect of DCK on enhancing the absorption of insulin resulted from transcellular processes as well as bile acid transporter activity.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)