Merck
Population pharmacokinetic analysis of piperacillin in burn patients.
Antimicrobial agents and chemotherapy (2014-04-23)
Sangil Jeon, Seunghoon Han, Jongtae Lee, Taegon Hong, Jeongki Paek, Heungjeong Woo, Dong-Seok Yim
ABSTRACT
Piperacillin in combination with tazobactam, a β-lactamase inhibitor, is a commonly used intravenous antibiotic for the empirical treatment of infection in intensive care patients, including burn patients. The purpose of this study was to develop a population pharmacokinetic (PK) model for piperacillin in burn patients and to predict the probability of target attainment (PTA) using MICs and concentrations simulated from the PK model. Fifty burn patients treated with piperacillin-tazobactam were enrolled. Piperacillin-tazobactam was administered via infusion for approximately 30 min at a dose of 4.5 g (4 g piperacillin and 0.5 g tazobactam) every 8 h. Blood samples were collected just prior to and at 1, 2, 3, 4, and 6 h after the end of the infusion at steady state. The population PK model of piperacillin was developed using NONMEM. A two-compartment first-order elimination PK model was finally chosen. The covariates included were creatinine clearance (CLCR), day after burn injury (DAI), and sepsis. The final PK parameters were clearance (liters/h) (equal to 16.6 × [CLCR/132] + DAI × [-0.0874]), central volume (liters) (equal to 25.3 + 14.8 × sepsis [0 for the absence or 1 for the presence of sepsis]), peripheral volume (liters) (equal to 16.1), and intercompartmental clearance (liters/h) (equal to 0.636). The clearance and volume of piperacillin were higher than those reported in patients without burns, and the terminal half-life and PTA decreased with the increased CLCR. Our PK model suggests that higher daily doses or longer durations of infusion of piperacillin should be considered, especially for burn patients with a CLCR of ≥ 160 ml/min.
Product Description
Piperacillin, European Pharmacopoeia (EP) Reference Standard
USP
Piperacillin, United States Pharmacopeia (USP) Reference Standard