Merck
  • Hemocompatibility evaluation of small elastomeric hollow fiber membranes as vascular substitutes.
Hemocompatibility evaluation of small elastomeric hollow fiber membranes as vascular substitutes.
Journal of biomaterials applications (2014-06-11)
Ángel E Mercado-Pagán, Dai Fei Elmer Ker, Yunzhi Yang
ABSTRACT
One of the main challenges for clinical implementation of small diameter vascular grafts (SDVGs) is their limited hemocompatibility. Important design specifications for such grafts include features that minimize the long-term risks of restenosis, fouling, and thrombus formation. In our lab, we have developed elastomeric hollow fiber membranes (HFMs), using a phase inversion method, as candidates for SDVGs. Here, we present our results for in vitro hemocompatibility testing of our HFM under flow and static conditions. Our results showed that the polymer-based HFMs do not damage the integrity of human red blood cells (RBCs) as shown by their low hemolytic extent (less than 2%). When analyzed for blood cell lysis using lactate dehydrogenase (LDH) activity as an indicator, no significant differences were observed between blood exposed to our HFMs and uncoagulated blood. Analysis of protein adsorption showed a low concentration of proteins deposited on the surfaces of HFM after 24 h. Platelet adhesion profiles using human platelet-rich plasma (PRP) showed that a low level of platelets adhered to the HFMs after 24 h, indicating minimal thrombotic potential. Under the majority of conditions, no significant differences were observed between medical-grade polymers and our HFMs. Eventual optimization of hemocompatible elastomeric HFM vessel grafts could lead to improved tissue vascularization as well as vascularized, tissue-engineered scaffolds for organ repair.
Sigma-Aldrich
Silica, mesostructured, SBA-15, 99% trace metals basis
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Silicon dioxide, purum p.a., acid purified
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Supelco
Glass spheres
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Glutaraldehyde solution, 25 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silicon dioxide, fused (granular), 1-3 mm, 99.9% trace metals basis
Sigma-Aldrich
Silicon dioxide, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Supelco
Silica, 99.8%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Silicon dioxide, acid washed and calcined, Analytical Reagent
Sigma-Aldrich
Silicon dioxide, granular, ≥99.9%
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Silicon dioxide, washed and calcined, analytical reagent
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Silicon dioxide, acid washed
Sigma-Aldrich
1-Naphthyl phosphate monosodium salt monohydrate, ≥98% (titration), powder
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use