Merck
  • Hydrophilic carbon nanoonions coated with carbon nanobubbles by laser ablation of carbon black suspension.
Hydrophilic carbon nanoonions coated with carbon nanobubbles by laser ablation of carbon black suspension.
Journal of nanoscience and nanotechnology (2013-07-19)
Fei Tian, Junchao An, Hongmei Cao, Shizhen Guo, Jing Zhao, Xiaozuo Lu
ABSTRACT
Hydrophilic carbon nanoonions coated with carbon nanobubbles were synthesized by long-pulse-width laser ablating of carbon black particles suspended in water. The high resolution TEM (HTREM), selected area electron diffraction (SAED) and Raman spectrometer results revealed that the crystallinity and the structure of carbon nanoonions. All analysis techniques showed that the hydrophilic properties of carbon nanoonions could be ascribed to both hydrophilic groups and carbon nanobubbles simultaneously formed on the surface of the synthesized carbon nanoonions under laser ablation, in which the carbon nanobubbles were also believed to improve the specific surface area and form more hydrophilic groups by chemical or physical absorption.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular