Merck
Comparability of portable nanoparticle exposure monitors.
The Annals of occupational hygiene (2012-07-04)
Christof Asbach, Heinz Kaminski, Daniel von Barany, Thomas A J Kuhlbusch, Christian Monz, Nico Dziurowitz, Johannes Pelzer, Katja Vossen, Knut Berlin, Silvio Dietrich, Uwe Götz, Heinz-Jürgen Kiesling, Rudolf Schierl, Dirk Dahmann
ABSTRACT
Five different portable instrument types to monitor exposure to nanoparticles were subject to an intensive intercomparison measurement campaign. Four of them were based on electrical diffusion charging to determine the number concentration or lung deposited surface area (LDSA) concentration of airborne particles. Three out of these four also determined the mean particle size. The fifth instrument type was a handheld condensation particle counter (CPC). The instruments were challenged with three different log-normally distributed test aerosols with modal diameters between 30 and 180 nm, varying in particle concentration and morphology. The CPCs showed the highest comparability with deviations on the order of only ±5%, independent of the particle sizes, but with a strictly limited upper number concentration. The diffusion charger-based instruments showed comparability on the order of ±30% for number concentration, LDSA concentration, and mean particle size, when the specified particle size range of the instruments matched the size range of the aerosol particles, whereas significant deviations were found when a large amount of particles exceeded the upper or lower detection limit. In one case the reported number concentration was even increased by a factor of 6.9 when the modal diameter of the test aerosol exceeded the specified upper limit of the instrument. A general dependence of the measurement accuracy of all devices on particle morphology was not detected.
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, ≥97.0% (GC)
Sigma-Aldrich
Bis(2-ethylhexyl) sebacate, technical grade, 90%
Supelco
Bis(2-ethylhexyl) sebacate, Selectophore, ≥97.0%