Skip to Content
Merck
Eshmuno® HCX Resin RoboColumn Eshmuno® HCX, volume 0.2 mL, prepacked column

Eshmuno® HCX Resin

Eshmuno® HCX Resin RoboColumn Eshmuno® HCX, volume 0.2 mL, prepacked column

Products

Product Category
Brand
Physical Form
Product Line
Analyte Suitability
Separation Technique
ID (mm)
Volume (ml)
Quality Segment
Application
Housing Material
Particle Size (µm)
Sterility