Skip to Content
Merck
All Photos(1)

Key Documents

1.25066

Millipore

Eshmuno® HCX Resin

MiniChrom Column Eshmuno® HCX, volume 1 mL, prepacked column

Synonym(s):

MiniChrom Column Eshmuno® HCX, 1 ml

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€176.00

€176.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
1 PKG
€176.00

About This Item

UNSPSC Code:
41115711

€176.00


Estimated to ship onMay 28, 2025


ligand

(Sulfo + carboxy + phenyl)

material

polypropylene housing

product line

Eshmuno®

form

prepacked column

parameter

≤8 bar pressure
1000 cm/hr max. flow rate

column I.D. × L

8 mm × 20 mm

volume

1 mL

matrix active group

Polyvinyl Ether

mean particle size

85 μm

capacity

~50 mL, resin binding capacity

application(s)

mAb
recombinant protein
vaccine development

separation technique

mixed mode

storage temp.

15-25°C

General description

8 x 20mm

Features and Benefits

Eshmuno® HCX chromatography resin enables:

  • Greater capacity at high-salt concentrations
  • Superior productivity
  • Outstanding selectivity
  • Rigid base beads for easy packing
  • Excellent pressure-flow behavior
  • Strong chemical stability

Analysis Note

Appearance: passes test

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service