Skip to Content
Merck
α-Chloralose ≥98.0% (chloralose, AT)

α-Chloralose

α-Chloralose ≥98.0% (chloralose, AT)
Synonym(s):
1,2-O-(2,2,2-Trichloroethylidene)-α-D-glucofuranose, Anhydro-D-glucochloral, Chloralose
Empirical Formula (Hill Notation):
C8H11Cl3O6
CAS Number:
Molecular Weight:
309.53
EC Number:

Products

Product Category
Brand
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing