V900826
Palladium(II) acetate
Vetec™, reagent grade, 98%
Synonym(s):
Pd(OAc)2, [Pd(OAc)2]3
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
Pd(OCOCH3)2
CAS Number:
Molecular Weight:
224.51
Beilstein:
6086766
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
Recommended Products
grade
reagent grade
Quality Level
product line
Vetec™
Assay
98%
mp
216.3-223.7 °C (dec.)
SMILES string
CC(O[Pd]OC(C)=O)=O
InChI
1S/2C2H4O2.Pd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2
InChI key
YJVFFLUZDVXJQI-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1A
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jinyue Ding et al.
Nature communications, 5, 5474-5474 (2014-11-19)
Non-aromatic nitrogen- and oxygen-containing heterocycles such as piperidines and pyrans are prevalent components of natural products and pharmaceutical drugs. Although it has been a workhorse as a synthetic method for assembling unsaturated sp(2)-hybridized substrates, transition metal-catalysed cross-coupling chemistry is traditionally
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service