Skip to Content
Merck
All Photos(1)

Key Documents

PS790

S-Bioallethrin

analytical standard

Synonym(s):

Esbiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
EC Number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-790

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C\C(C)=C\[C@@H]1[C@@H](C(=O)O[C@H]2CC(=O)C(CC=C)=C2C)C1(C)C

InChI

1S/C19H26O3/c1-7-8-13-12(4)16(10-15(13)20)22-18(21)17-14(9-11(2)3)19(17,5)6/h7,9,14,16-17H,1,8,10H2,2-6H3

InChI key

ZCVAOQKBXKSDMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F

Flash Point(C)

120 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Liu et al.
Nan fang yi ke da xue xue bao = Journal of Southern Medical University, 26(3), 321-324 (2006-03-21)
To study the effect of the S-bioallethrin on human lymphocytes by microarray technique. The changes of normal human lymphocytes treated with S-bioallethrin were examined with light microscope, flow cytometry, electron microscope, DNA ladder and microarray techniques. Morphological study showed that
J Ahlbom et al.
Brain research, 645(1-2), 318-324 (1994-05-09)
This study shows that neonatal exposure to the insecticide bioallethrin has a dose-dependent effect on muscarinic cholinergic receptors (MAChR) in the neonatal mouse, leading to permanent changes in MAChR and in spontaneous behaviour in adult mice. Neonatal NMRI mice, given
S Sulaiman et al.
Journal of vector ecology : journal of the Society for Vector Ecology, 25(1), 94-97 (2000-08-05)
Deltacide (S-bioallethrin 0.71% w/v, deltamethrin 0.5% w/v, piperonyl butoxide 8.9% w/v excipients to 100% w/v) and Solfac UL 015 (cyfluthrin 1.5% w/v) were evaluated against the sentinel sugar-fed adults and 4th-instar larvae of Aedes aegypti at 17 storey high-rise apartments
Z Q Zhou et al.
Se pu = Chinese journal of chromatography, 19(6), 526-528 (2003-01-28)
A chiral stationary phase(CSP) was prepared by coating cellulose-tris (3,5-dimethylphenylcarbamate) onto aminopropylated spherical silica gel. On the CSP, the chiral separation of the bioallethrin enantiomers has been investigated and under the optimum conditions the optical purity of three samples of
Pyrethroids and the synergist piperonyl-butoxide (PBO) affect T-cells and basophils.
F Diel et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 48 Suppl 1, S15-S16 (1999-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service