Skip to Content
Merck
All Photos(1)

Documents

PS47

Silvex®

analytical standard

Synonym(s):

2-(2,4,5-Trichlorophenoxy)propionic acid, 2,4,5-TP, Fenoprop, Silvex®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7Cl3O3
CAS Number:
Molecular Weight:
269.51
Beilstein:
1985768
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-47

application(s)

agriculture
environmental

SMILES string

CC(Oc1cc(Cl)c(Cl)cc1Cl)C(O)=O

InChI

1S/C9H7Cl3O3/c1-4(9(13)14)15-8-3-6(11)5(10)2-7(8)12/h2-4H,1H3,(H,13,14)

InChI key

ZLSWBLPERHFHIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silvex®(2-(2,4,5-trichlorophenoxy)-propionic acid) is a chlorophenoxy acid pesticide. It is used to control annual and perennial broadleaf weeds.

Application

2-(2,4,5-Trichlorophenoxy)propionic acid may be used as a reference standard for the determination of Silvex® (2-(2,4,5-Trichlorophenoxy)propionic acid), in soil and water samples by enzyme-linked immunosorbent assay (ELISA).

Legal Information

Silvex is a registered trademark of Silberline Manufacturing Co., Inc.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergi Morais et al.
Analytical and bioanalytical chemistry, 374(2), 262-268 (2002-09-27)
A highly sensitive and specific indirect enzyme-linked immunosorbent assay is described for Silvex, 2-(2,4,5 trichlorophenoxy)propionic acid, (2,4,5-TP). One specific feature of the immunoassay is the use of simple chemical activation of chlorophenoxy acids to prepare both the immunizing and coating
Emeline L Maillet et al.
Journal of medicinal chemistry, 52(21), 6931-6935 (2009-10-13)
We show that phenoxyauxin herbicides and lipid-lowering fibrates inhibit human but not rodent T1R3. T1R3 as a coreceptor in taste cells responds to sweet compounds and amino acids; in endocrine cells of gut and pancreas T1R3 contributes to glucose sensing.
Lourdes Santana et al.
Journal of medicinal chemistry, 51(21), 6740-6751 (2008-10-07)
The work provides a new model for the prediction of the MAO-A and -B inhibitor activity by the use of combined complex networks and QSAR methodologies. On the basis of the obtained model, we prepared and assayed 33 coumarin derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service