Skip to Content
Merck
All Photos(1)

Documents

NEKIT420

Ethera NEMo Portable Indoor Air Quality (IAQ) Logger

Synonym(s):

Profil`Air®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

availability

available only in EU

size

29 × 21.5 × 10

application(s)

environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

Formaldehyde is one of the main pollutants of indoor air. This carcinogenic product is present in many manufactured goods used in building construction or decorative products. It is ubiquitous in dwellings and offices, wherre we spend over 80 % of our time. In the long term, formaldehyde can lead to serious health problems, especially affecting respiratory tract diseases (asthma, bronchial irritations . . . ).

Features and Benefits

Portable logger of Indoor Air Quality (IAQ) to continuously and selectively measure confinement and formaldehyde with the levels stipulated for indoor air quality.

Suitability

Fits the following instruments:
NEFOR011

Other Notes

Weight: 0.7
Temperature conditions: Room temperature

Legal Information

Profil'Air is a registered trademark of Ethera S.A.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service