G1007604
GC/MS/MS Analysis of Pesticides in Chamomile on SLB®-5ms after QuEChERS Cleanup using SupelClean™ Ultra
suitable for GC/MS
About This Item
Recommended Products
technique(s)
GC/MS: suitable
test parameters
sample/matrix: chamomile
extraction tube: Supel™ Que non-buffered extraction tube (4 mg NaSO4, 1 g NaCl in 12 mL tube) (55294-U)
extraction process: Add 10 mL acetonitrile/IS Extraction Solvent and allow to sit 15 minutes, then shake or vortex 30 sec. Add contents of Supel Que non-buffered extraction tube (55294-U). (Shake by hand immediately to prevent clumping. Shake on shaker for 30 minutes at 2250 rpm, centrifuge for 5 minutes at 4500 rpm. Draw off supernatant for cleanup.)
clean-up tube: Supelclean Ultra 2400, 3 mL (54281-U)
clean-up process: See description text
column: SLB-5ms, 30 m x 0.25 mm I.D. x 0.25 μm (28471-U)
oven: 105°C (3 min), 10 °C/min to 130 °C, 4°C/min to 200°C, 8°C/min. to 300°C (10 min)
inj. temp.: programmed: 100 °C (.05 min), to 280 °C at 720°C/min, hold 45 min
detector: MS/MS
MSD interface: 300 °C
scan range: MRM
carrier gas: helium, 1.4 mL/min constant flow
injection: 2 μL splitless
liner: 2 mm ID
sample: QuEChERS extract of chamomile
application(s)
food and beverages
Analysis Note
Other Notes
Cartridge: Top 3 mL Supelclean Ultra 2400 SPE cartridge (54281-U) with ~0.10 g anhydrous sodium sulfate
Condition: 3 mL of acetonitrile
Load: 750 μL of QuEChERS extract
Elute: using 8 mL of acetonitrile containing 0.5% formic acid, apply slight vacuum at end if needed.
Collect eluent in glass or plastic centrifuge tubes (glass preferred)
Concentrate: all eluent extracts in water bath at 45-50°C w/3 psi nitrogen purge to ~50-100 μL.
Legal Information
Analyte
accessory
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service