Skip to Content
Merck
All Photos(1)

Key Documents

G1006858

LC/MS Analysis of Nucleosides on Ascentis® Express F5

application for LC-MS

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

technique(s)

LC/MS: suitable

test parameters

column: Ascentis Express F5, 15 cm x 2.1 mm I.D., 2.7 μm particles, 90Å (53571-U)
mobile phase: [A] 5 mM ammonium acetate, pH 5.5 with acidic acid in water; [B] Methanol
gradient: hold at 0% for 6 min; to 50% B in 4 min; hold at 50% B for 4 min; to 0% B in 0.1 min; to hold at 0% B for 5.9 min
flow rate: 0.3 mL/min
pressure: 5017-9367 psi
column temp.: 15 °C
detector: MS, ESI (-) QTOF, EIC
injection: 5 μL
sample: 100 μg/mL of 12 nucleosides except N6-methyladenosine in 99:1 10mM ammonium acetate pH 5.5 in water : methanol

suitability

application for LC-MS

application(s)

life science and biopharma

General description

Detector: MS, ESI (-) QTOF, EIC, (Pseudouridine m/z 243 and 153; Cytidine m/z 242 and 109; Uridine m/z 243 and 111; 2-Thiocytidine m/z 258 and 126; Guanosine m/z 282 and 150; Inosine m/z 267 and 135; Ribothymidine m/z 257 and 125; 1-Methyladenosine m/z 280 and 148; 2′-O-Methyladenosine m/z 256, 213 and 108; 5-Methylcytidine m/z 256 and 124; 7-Methylguanosine m/z 124 and 164; 3-Methylcytidine methosulfate m/z 256 and 124; N6-Methyladenosine m/z 280 and 148.

Analysis Note

Nucleosides are structural subunits of nucleic acids, the macromolecules that convey genetic information in living cells. They contain a molecule of sugar (pentose) bonded to a nitrogen-containing organic ring compound. The nitrogen-containing compound is either a pyrimidine (cytosine, thymine, or uracil) or a purine (adenine or guanine). Nucleosides, derivatives and / or analogues can be used in many different industries and applications as in agrochemistry (herbicides, fungicides and insecticides), therapeutic drugs including a range of antiviral (HIV, hepatitis C virus and herpes simplex) and even antibiotics. This application demonstrates reverse phase interactions with mobile phases that are shown to work well in mass spectrometry conditions.

Other Notes

1-methyladenosine undergoes a Dimroth rearrangement to yield N6-methyladenosine in the standard solution mix.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Pseudouridine
    Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
    Uridine, ≥99%
    2-Thiocytidine
    5-Methylcytidine, ≥99%
    Ribothymidine
    2′-O-Methylcytidine
    Inosine, ≥99% (HPLC)
    Guanosine, ≥98%
    3-Methylcytidine methosulfate
    1-Methyladenosine
    7-Methylguanosine, ≥90%
    N6-Methyladenosine
    See All (13)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service