G005942
HPLC Analysis of mAb on Antibodix® WCX-NP5, 25 cm Columns
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: Antibodix WCX-NP5, 25 cm x 4.6 mm I.D., 5 μm particles (Z777294)
mobile phase: (A) 10 mM sodium phosphate buffer, pH 7.5; (B) A + 100 mM NaCl
gradient: 15 to 55% B in 30 min
flow rate: 0.8 mL/min
column temp.: ambient
detector: UV, 214 nm
injection: 10 μL
sample: monoclonal antibody
suitability
application for HPLC
application(s)
life science and biopharma
pharmaceutical (small molecule)
Legal Information
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service