Skip to Content
Merck
All Photos(1)

Documents

APP501

HPLC Analysis of Cinacalcet Hydrochloride Impurities with a Purospher® STAR Phenyl Column

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Purospher® STAR Phenyl (5 µm) LiChroCART® 150 x 4.6 mm I.D. (1.51922)
mobile phase: [A] 10 mmol/L Sodium perchlorate solution, adjusted to pH 2.5 with perchloric acid; [B] Acetonitrile
gradient: 0-0.5 min 2.5% B, in 4.5 min to 95%B, 2 min 95%B, 3 min at 2.5%B
flow rate: 1.0 mL/min
pressure: 60 bar (870 psi)
column temp.: 60°C (Sampler: 10 °C)
detector: UV, 215 nm (analytical flow cell; 10 µL)
injection: 5 µL
sample/matrix: Impurity identification solution with cinacalcet impurities A, B, C at 1.5 µg/mL each and cinacalcet at 5 µg/mL in solvnt mixture (10 mmol/L sodium perchlorate solution, adjusted to pH 2.5 with perchloric acid, and acetonitrile in a ratio of 50:50 (%v/v))

application(s)

pharmaceutical

General description

Cinacalcet hydrochloride is an oral calcimimetic indicated for the treatment of secondary hyperthyroidism in patients on dialysis with end-stage renal disease, and in patients with parathyroid carcinoma to reduce hypercalcemia.

Analysis Note

The developed method on a Purospher® STAR Phenyl, 150 × 4.6 mm (5µm) column was found to be linear for cinacalcet hydrochloride up to a concentration of 6 µg/mL with a LOQ of 0.65 µg/mL and LOD of 0.21 µg/mL. For cinacalcet impurity A, impurity B and impurity C, the method was linear up to a concentration of 1.8 µg/mL with LOQ ranging from 0.03 to 0.13 µg/mL and LOD ranging from 0.01 to 0.04 µg/mL. The method can be applied for the study of related substances in cinacalcet hydrochloride API using the Purospher® STAR Phenyl HPLC column. Repeatability for the cinacalcet 5 µg/mL solution was 1.19% RSD (n=5). The tailing factor for cinacalcet eluting at 12.3 min was NMT 2.0 (1.31) and the theoretical plate count was significantly above 2000.

Other Notes

App_501

Legal Information

LICHROCART is a registered trademark of Merck KGaA, Darmstadt, Germany
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a stability-indicating RP-UPLC method for the estimation of impurities in cinacalcet hydrochloride API and its formulation
Reddy PS, et al.
Scientia Pharmaceutica, 83(4) (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service