Skip to Content
Merck
All Photos(1)

Documents

821488

TSKgel® SuperMultiporeHZ-M (4 µm) HPLC Columns

L x I.D. 15 cm × 4.6 mm, polystyrene-divinylbenzene phase

Synonym(s):

TSKgel® Size Exclusion (H-Type) HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® SuperMultiporeHZ-M HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 15 cm × 4.6 mm, 4 μm particle size

material

stainless steel hardware

Agency

suitable for USP L21

product line

TSKgel®

manufacturer/tradename

Tosoh 21488

parameter

0.4 mL/min flow rate
24 bar max. pressure
60 °C temperature

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

polymer particle platform

matrix active group

polystyrene-divinylbenzene phase

particle size

4 μm

pore size

mixed

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

The innovative multi-pore technology is now also available in 3, 4 and 6 micron TSKgel SuperMultiporeHZ columns. Three columns, varying in linear range, are available. The base material of each TSKgel SuperMultiporeHZ column is poly(styrene/divinylbenzene). Particles in TSKgel SuperMultiporeHZ columns are mono-disperse in size and exhibit a broad range of pore sizes. Each particle, by design, has an extended linear calibration curve, thereby greatly diminishing chromatograms with inflection points. GPC chromatograms without inflection points . . . it will take some getting used to.
The TSKgel HXL column line consists of eight columns with different pore sizes, TSKgel G1000HXL through TSKgel G7000HXL, and three columns with an extended linear range of the calibration curve, TSKgel GMHXL , TSKgel GMHXL-L and TSKgel MultiporeHXL-M. Main characteristics are: ultra-low sample adsorption, i.e., the columns show true size exclusion behavior for most polymers, maximum operating temperature for TSKgel Hxl-type columns is 60°C for G1000HXL , G2000HXL , G2500HXL and G3000HXL , and 80°C for the remaining columns in the product line. See TSKgel HHR HT columns for operating temperatures of 140°C, the columns are shipped in THF, which can be exchanged for a limited number of organic solvents. TSKgel HXL-type columns are conventional GPC columns with column dimensions of 7.8mm ID x 30cm.

Physical form

Column can only be used in THF solvent.

Other Notes

Uses guard 821489, available as a custom product. Please inquire.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service