Skip to Content
Merck
All Photos(1)

Key Documents

808651

TOYOPEARL® AF-Red-650M

bottle of 25 mL, 65 μm particle size

Synonym(s):

TOYOPEARL® AF-Red-650M Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€205.00

€205.00


Check Cart for Availability


Select a Size

Change View
25 ML
€205.00

About This Item

UNSPSC Code:
41115700

€205.00


Check Cart for Availability

description

Adsorption capacity (3.5 +/- 1.0 mg/mL (human serum albumin))
Phase filter (Red)
Shipping solvent (20% (v/v) ethanol)

product line

TOYOPEARL®

form

slurry

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh 08651

extent of labeling

7 μmol/mL ligand density

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

Procion Red HE-3B phase

particle size

65 μm

pore size

1000 Å

pH

4-9

capacity

3.5 g/L±1.0 adsorption capacity (human serum albumin)

separation technique

affinity

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl AF-Red-650 resin is functionalized with Procion Red HE-3B (also known as Reactive Red 120). It is used for nucleotide-dependent enzymes, lipoproteins, plasminogen, peptides, hormones, and cytotoxins

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service