Skip to Content
Merck
All Photos(2)

Documents

794-0399

GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean ENVI-8

suitable for GC, application for SPE

Synonym(s):

Chlorinated pesticides in hazardous waste

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

test parameters

sample preparation: SPE (Solid Phase Extraction)
sample/matrix: waste water spiked with pesticides
SPE tube/cartridge: Supelclean ENVI-8, glass hardware, 500 mg/6 mL (57107 (500 mg/6 mL); 57108 (1 g/6 mL))
condition: 3 mL methanol; h 2 mL 5% methanol in water
sample addition: load sample at 5 mL/min
drying: 2-3 min with nitrogen stream
elution: 2 x 4 mL hexane:acetone (90:10), drop wise. allow to soak into bed
column: 5% Phenyl Polysiloxane, 15 m × 0.53 mm I.D., 0.50 μm (Supelco equivalent, Equity-5, 28252-U)
oven: 150 °C (0.5 min) to 275 °C (5 min)
inj. temp.: 200 °C
detector: ECD, 300 °C
carrier gas: helium
injection: 1 μL split/splitless (45 sec delay)

suitability

application for SPE

application(s)

environmental
food and beverages

Application

Sample Pre-treatment:
Add 5 mL methanol to 100 mL aqueous hazardous waste. Adjust to pH 5-7 if necessary.

Legal Information

ENVI is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    Aldrin
    Endrin
    γ-BHC, certified reference material, 1000 μg/mL in methanol
    Endosulfan sulfate
    Methoxychlor
    α-BHC
    4,4′-DDD, PESTANAL®, analytical standard
    γ-Chlordane
    Decachlorobiphenyl
    Heptachlor
    4,4′-DDT
    Endrin aldehyde
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene
    Lindane
    Endrin ketone
    β-BHC
    Heptachlor epoxide, PESTANAL®, analytical standard, isomer B
    δ-BHC
    Dieldrin
    α-Chlordane
    See All (20)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service