Skip to Content
Merck
All Photos(1)

Documents

58695-U

FEP Tubing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32119290
NACRES:
SB.52

parameter

50 °C max. temp.
500 psi max. pressure (35 kg)

Looking for similar products? Visit Product Comparison Guide

General description

For connecting pump to reservoirs. Use 1/8 in. O.D. × 0.0625 in. I.D. tubing with most pumps, 0.15 in. O.D. × 0.118 in. I.D. tubing with Waters pumps. 10 ft./3 m length.
PTFE FEP tubing is ideal for use at pressures up to 500 psi (35 kg/cm2).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service