Skip to Content
Merck
All Photos(1)

Documents

57254

Supelco

Visiprep Manifold Replacement Gasket

for use with 24-port model (standard and DL), pk of 2

Synonym(s):

REPL. GASKET PK/2 FOR 24 PORT VACUUM MANIFOLD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

packaging

pk of 2

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with 24-port model (standard and DL)

Looking for similar products? Visit Product Comparison Guide

General description

Visiprep Manifold Replacement Gasket is an accessory for use with 24-port Visiprep solid-phase extraction manifold.

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service