57187
PTFE Frit, 20 μm porosity
for use with 6mL SPE tubes, pk of 60
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
for SPE Tube
packaging
pk of 60
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with 6mL SPE tubes
Looking for similar products? Visit Product Comparison Guide
General description
Frits are important components of the high performance liquid chromatography system design and is employed as a depth filter for the concurrent physical removal of small molecules. Frits are available in various materials. Medium, 20 μm porosity frits are used for crystalline precipitates and extraction applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service