51395-U
Upchurch PEEK Fingertight Fitting
long 2 piece
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
Recommended Products
packaging
pack of 10
I.D.
0.0625 in. (1.5875 mm)
Looking for similar products? Visit Product Comparison Guide
General description
The one-piece fitting, 55067-U, is convenient to use because the ferrule will not stick in a receiving port and can be easliy found if dropped onto the floor. The two-piece fitting features a separate ferrule so you can replace just the ferrule instead of the entire unit. The Upchurch PEEK Fingertight fittings are for use with 1/16″ O.D. tubing and can be used to 6,000 psi (420 kg/cm2).
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service