Skip to Content
Merck
All Photos(1)

Key Documents

47829

Supelco

D-(+) Glucose

analytical standard

Synonym(s):

D-(+)-Glucose, Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

150-152 °C (lit.)

solubility

H2O: soluble

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Product 47829 has been discontinued. Product 47249 may be an acceptable replacement.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dextrose as carbon source in the culture of Litopenaeus vannamei (Boone, 1931) in a zero exchange system
Suita.MS, et al.
Latin American Journal of Aquatic Research, 43(3), 2017-2017 null
NPCS board et al.
Investment Opportunity in Drugs & Pharmaceutical Projects (2013)
Yu-ting Alex Chiang et al.
American journal of physiology. Endocrinology and metabolism, 306(7), E707-E722 (2013-12-26)
p21-Activated protein kinases (PAKs) are centrally involved in a plethora of cellular processes and functions. Their function as effectors of small GTPases Rac1 and Cdc42 has been extensively studied during the past two decades, particularly in the realms of cell
Lars P Bechmann et al.
Journal of hepatology, 56(4), 952-964 (2011-12-17)
It is widely known that the liver is a central organ in lipogenesis, gluconeogenesis and cholesterol metabolism. However, over the last decades, a variety of pathological conditions highlighted the importance of metabolic functions within the diseased liver. As observed in
Dov B Ballak et al.
Nature communications, 5, 4711-4711 (2014-09-04)
Cytokines of the IL-1 family are important modulators of obesity-induced inflammation and the development of systemic insulin resistance. Here we show that IL-1 family member IL-37, recently characterized as an anti-inflammatory cytokine, ameliorates obesity-induced inflammation and insulin resistance. Mice transgenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service