Skip to Content
Merck
All Photos(1)

Key Documents

442721

Supelco

o-Terphenyl

analytical standard

Synonym(s):

1,2-Diphenylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4C6H5
CAS Number:
Molecular Weight:
230.30
Beilstein:
1908446
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

337 °C (lit.)

mp

56-59 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

c1ccc(cc1)-c2ccccc2-c3ccccc3

InChI

1S/C18H14/c1-3-9-15(10-4-1)17-13-7-8-14-18(17)16-11-5-2-6-12-16/h1-14H

InChI key

OIAQMFOKAXHPNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

325.4 °F

Flash Point(C)

163 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keewook Paeng et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(16), 4952-4957 (2015-04-01)
The concept of dynamic heterogeneity and the picture of the supercooled liquid as a mosaic of environments with distinct dynamics that interchange in time have been invoked to explain the nonexponential relaxations measured in these systems. The spatial extent and
Joaquim Vila et al.
Applied and environmental microbiology, 75(19), 6232-6239 (2009-08-12)
The pyrene-degrading Mycobacterium sp. strain AP1 grew in nutrient-supplemented artificial seawater with a heavy fuel oil as the sole carbon source, causing the complete removal of all linear (C(12) to C(40)) and branched alkanes from the aliphatic fraction, as well
Shijia Liu et al.
Analytical and bioanalytical chemistry, 407(30), 8989-8997 (2015-10-03)
A novel imidazolium-embedded N,N-dimethylaminopropyl-functionalized silica-based stationary phase (Sil-ImCl) was prepared and further used for hydrophilic interaction/reversed-phase mixed-mode chromatography. The Sil-ImCl stationary phase was respectively characterized by Fourier transform infrared spectrometry, thermogravimetric analysis, and element analysis. A variety of hydrophilic or
Robert Amdany et al.
Environmental monitoring and assessment, 186(9), 5639-5653 (2014-05-30)
In this study, the semipermeable membrane device (SPMD) passive samplers were used to determine freely dissolved concentrations of polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs) and organochlorine pesticides (OCPs) in selected water bodies situated in and around Johannesburg City, South
Michał Szumski et al.
Journal of chromatography. A, 1373, 114-123 (2014-12-04)
A novel, cholesterol-based polymeric monolithic stationary phase for capillary liquid chromatography, was prepared by thermally initiated in-situ polymerization. Cholesteryl methacrylate (CholMA) was used as a functional monomer and trimethylolpropane trimethacrylate (TRIM) was a cross-linker, while azobisisobutyronitrile (AIBN) was an initiator.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service