23204-U
Space Saver storage racks
for use with 6-8 mm diamerter vial (96 vials per rack), well opening 9.5 mm, pkg of 5 ea
Synonym(s):
VIAL RACK,8MM VIALS, 5/CS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
red polypropylene rack
feature
positions 96 (vials per rack)
packaging
pkg of 5 ea
rack L × W × H
183 mm × 125 mm × 23 mm
well opening
9.5 mm
compatibility
for use with 6-8 mm diamerter vial (96 vials per rack)
Looking for similar products? Visit Product Comparison Guide
General description
Economical and convenient storage for samples in your screw cap and crimp seal vials. Can be stacked when empty or full. Autoclavable, 120°C. Each case contains 5 racks.
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service