Z740356
Nunc® petri dishes
H 150 mm × 20 mm, surface area × working volume 145 cm2 × 35 mL, lid
Synonym(s):
culture dishes, disposable petri dishes, plastic petri dishes
All Photos(1)
About This Item
Recommended Products
material
clear polystyrene
sterility
sterile
feature
lid
packaging
pack of 80 ea
manufacturer/tradename
Nunc 168381
H
150 mm × 20 mm
dish diam. × H
150 mm × 20 mm
surface area × working volume
145 cm2 × 35 mL
binding type
Nunclon Delta surface
General description
Polystyrene provides high optical quality-especially when microscopic examination is required.
Includes:
Includes:
- A certificate which assures user of biological performance as well as traceability
- Top and bottom stacking rings for easier handling and more stable stacking
- Tested for cell growth and plating efficiency
- Radiation sterilized
- Air vents provide gas exchange
- For cloning experiments and plating efficiency determinations
- With lid
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries