Key Documents
Z645451
Nichiryo pipette tips
maximum volume 1000 μL, universal, bulk pack
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
Pricing and availability is not currently available.
material
clear tip
colorless
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (Bag)
manufacturer/tradename
Nichiryo BMT-L(B)
availability
not available in (France)
maximum volume
1000 μL
Features and Benefits
  • RNase and DNase free
  • Fits most pipettes
  • Premium packaging featuring stand-up and puncture-resistant bags
  • Racked tips are offered in both hinged and lift-off tops
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)