Documents
Z641987
Hettich® centrifuge adapters
for ROTINA 38/38R, ROTINA 420/420R and ROTANTA 460/460R Centrifuges, Adapter for 8.5-15 mL tubes (16-17x100 mm)
About This Item
UNSPSC Code:
41103911
NACRES:
NB.22
feature
Holds 8.5 mL tubes
packaging
pack of 8 ea
manufacturer/tradename
Hettich 5248
technique(s)
centrifugation: suitable
compatibility
for use with 5051 (bucket )
General description
  • compatible with bucket/carrier 5051and 5628
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG